Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100331.1 | NP_001093801.1 | 9 - 10 | 1044 | 68 | |
GenBank mRNA | BC149261.1 | - | 9 - 10 | 1044 | 68 | |
KC787698.1 | - | 9 - 10 | 1043 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |