Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100342.2 | NP_001093812.1 | 2 - 3 | 631 | 101 | |
GenBank mRNA | BC149854.1 | - | 2 - 3 | 627 | 101 | |
BC149875.1 | - | 2 - 3 | 635 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |