Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100354.1 | NP_001093824.1 | 6 - 7 | 1546 | 77 | |
GenBank mRNA | BC147922.1 | - | 6 - 7 | 1546 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |