Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100381.1 | NP_001093851.1 | 4 - 5 | 476 | 115 | |
GenBank mRNA | BC150134.1 | - | 4 - 5 | 476 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |