Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101992.2 | NP_001095462.1 | 6 - 7 | 914 | 63 | |
GenBank mRNA | BC149291.1 | - | 6 - 7 | 915 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |