Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUBCNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102041.2 | NP_001095511.1 | 11 - 12 | 1771 | 102 | |
GenBank mRNA | BC151662.1 | - | 11 - 12 | 1772 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |