Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105200.1 | NP_001098670.1 | 8 - 9 | 1056 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |