Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105364.1 | NP_001098834.1 | 3 - 4 | 243 | 129 | |
GenBank mRNA | BC134427.1 | - | 3 - 4 | 243 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |