Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110074.1 | NP_001103544.1 | 2 - 3 | 218 | 151 | |
GenBank mRNA | BC108161.1 | - | 2 - 3 | 218 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |