Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110077.2 | NP_001103547.1 | 1 - 2 | 81 | 103 | |
GenBank mRNA | BC149287.1 | - | 1 - 2 | 82 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |