Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110181.1 | NP_001103651.1 | 10 - 11 | 1069 | 92 | |
GenBank mRNA | BT021196.1 | - | 10 - 11 | 1069 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |