Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 8 - 9 | 741 | 108 | |
GenBank mRNA | BC105446.1 | - | 8 - 9 | 777 | 108 | |
DV883546.1 | - | 5 - 6 | 423 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |