Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078099.2 | NP_001071567.2 | 1 - 2 | 336 | 73 | |
GenBank mRNA | BC123671.1 | - | 1 - 2 | 355 | 73 | |
BT030476.1 | - | 1 - 2 | 336 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |