Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098126.2 | NP_001091595.1 | 2 - 3 | 389 | 115 | |
GenBank mRNA | BC140614.1 | - | 2 - 3 | 390 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |