Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098146.1 | NP_001091615.1 | 4 - 5 | 347 | 71 | |
GenBank mRNA | BC140589.1 | - | 4 - 5 | 347 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |