Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099064.1 | NP_001092534.1 | 8 - 9 | 1308 | 70 | |
GenBank mRNA | BC142531.1 | - | 8 - 9 | 1308 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |