Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099108.2 | NP_001092578.1 | 5 - 6 | 449 | 73 | |
GenBank mRNA | BC146137.1 | - | 5 - 6 | 450 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |