Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099136.1 | NP_001092606.1 | 1 - 2 | 106 | 75 | |
GenBank mRNA | BC142148.1 | - | 1 - 2 | 106 | 75 | |
HM921049.1 | - | 1 - 2 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |