Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101206.1 | NP_001094676.1 | 6 - 7 | 613 | 70 | |
GenBank mRNA | BC147916.1 | - | 6 - 7 | 613 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |