Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101270.1 | NP_001094740.1 | 3 - 4 | 356 | 69 | |
GenBank mRNA | BC147901.1 | - | 3 - 4 | 356 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |