Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101274.2 | NP_001094744.1 | 5 - 6 | 422 | 71 | |
GenBank mRNA | BC149691.1 | - | 5 - 6 | 423 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |