Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC85A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102095.1 | NP_001095565.1 | 1 - 2 | 525 | 102 | |
GenBank mRNA | BC148154.1 | - | 1 - 2 | 525 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |