Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102136.1 | NP_001095606.1 | 5 - 6 | 897 | 57 | |
GenBank mRNA | BC148092.1 | - | 5 - 6 | 897 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |