Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102167.1 | NP_001095637.1 | 9 - 10 | 3452 | 91 | |
GenBank mRNA | BC148040.1 | - | 9 - 10 | 3452 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |