Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102244.2 | NP_001095714.1 | 4 - 5 | 420 | 105 | |
GenBank mRNA | BC149011.1 | - | 4 - 5 | 422 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |