Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102247.1 | NP_001095717.1 | 2 - 3 | 327 | 86 | |
GenBank mRNA | BC151363.1 | - | 2 - 3 | 327 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |