Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102277.1 | NP_001095747.1 | 9 - 10 | 1063 | 82 | |
GenBank mRNA | BC149831.1 | - | 9 - 10 | 1063 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |