Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102301.2 | NP_001095771.1 | 2 - 3 | 303 | 124 | |
GenBank mRNA | BC149380.1 | - | 2 - 3 | 304 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |