Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102320.1 | NP_001095790.1 | 4 - 5 | 1248 | 72 | |
GenBank mRNA | BC149093.1 | - | 4 - 5 | 1248 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |