Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104993.2 | NP_001098463.1 | 2 - 3 | 773 | 119 | |
GenBank mRNA | BC149902.1 | - | 2 - 3 | 774 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |