Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104996.2 | NP_001098466.1 | 1 - 2 | 431 | 79 | |
GenBank mRNA | BC133453.1 | - | 1 - 2 | 432 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |