Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109984.1 | NP_001103454.1 | 3 - 4 | 295 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |