Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111108.2 | NP_001104578.1 | 3 - 4 | 514 | 64 | |
NM_001111109.2 | NP_001104579.1 | 2 - 3 | 763 | 64 | ||
GenBank mRNA | EF446904.1 | - | 2 - 3 | 763 | 64 | |
EF446905.1 | - | 3 - 4 | 514 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |