Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101282.1 | NP_001094752.1 | 4 - 5 | 987 | 136 | |
GenBank mRNA | BC147990.1 | - | 4 - 5 | 987 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |