Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102353.1 | NP_001095823.1 | 5 - 6 | 847 | 57 | |
GenBank mRNA | BC112828.1 | - | 5 - 6 | 782 | 57 | |
BC149974.1 | - | 5 - 6 | 847 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |