Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103332.2 | NP_001096802.1 | 1 - 2 | 41 | 65 | |
GenBank mRNA | BC149748.1 | - | 1 - 2 | 42 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |