Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105029.1 | NP_001098499.1 | 15 - 16 | 1389 | 71 | |
GenBank mRNA | BC140677.1 | - | 15 - 16 | 1389 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |