Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105462.1 | NP_001098932.1 | 12 - 13 | 1291 | 80 | |
GenBank mRNA | BC134487.1 | - | 12 - 13 | 1291 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |