Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105486.2 | NP_001098956.1 | 2 - 3 | 277 | 84 | |
GenBank mRNA | BC149277.1 | - | 2 - 3 | 280 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |