Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176627.1 | NP_788800.1 | 8 - 9 | 994 | 141 | |
GenBank mRNA | AB098761.1 | - | 1 - 2 | 297 | 141 | |
AB098809.1 | - | 1 - 2 | 141 | |||
AB098814.1 | - | 443 | 141 | |||
AB099049.1 | - | 4 - 5 | 427 | 141 | ||
AF192334.1 | - | 8 - 9 | 994 | 141 | ||
BC134714.1 | - | 8 - 9 | 141 | |||
BC151448.1 | - | 8 - 9 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |