Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034788.2 | NP_001029960.2 | 5 | 957 | 132 | |
GenBank mRNA | BC112468.1 | - | 5 | 957 | 132 | |
BT021663.1 | - | 5 | 719 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Both primers and probe map within a single exon |