Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075939.2 | NP_001069407.1 | 1 | 154 | 142 | |
GenBank mRNA | BC120041.1 | - | 1 | 155 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |