Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079612.1 | NP_001073080.1 | 1 | 1750 | 143 | |
GenBank mRNA | BC122672.1 | - | 1 | 1750 | 143 | |
BC150113.1 | - | 3 | 2469 | 143 | ||
BC151579.1 | - | 1 | 2122 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Both primers and probe map within a single exon |