Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099371.1 | NP_001092841.1 | 1 | 1444 | 84 | |
GenBank mRNA | BC142154.1 | - | 1 | 1444 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |