Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174813.2 | NP_777238.1 | 2 | 941 | 148 | |
GenBank mRNA | M34336.1 | - | 2 | 941 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |