Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105324.2 | NP_001098794.1 | 6 - 7 | 749 | 72 | |
GenBank mRNA | BC133441.1 | - | 6 - 7 | 751 | 72 | |
BC149594.1 | - | 6 - 7 | 764 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |