Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105505.1 | NP_001098975.1 | 4 - 5 | 388 | 78 | |
GenBank mRNA | BC149353.1 | - | 4 - 5 | 388 | 78 | |
BT029884.1 | - | 4 - 5 | 380 | 78 | ||
BT029908.1 | - | 4 - 5 | 380 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |