Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128508.2 | NP_001121980.1 | 1 - 2 | 267 | 84 | |
GenBank mRNA | BC109877.1 | - | 1 - 2 | 268 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |