Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161651.1 | NP_001155123.1 | 5 - 6 | 562 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |