Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205470.1 | NP_001192399.1 | 3 - 4 | 408 | 75 | |
GenBank mRNA | BC151667.1 | - | 3 - 4 | 381 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |